MilliporeSigma
  • A simple and reliable pretreatment protocol facilitates fluorescent in situ hybridisation on tissue microarrays of paraffin wax embedded tumour samples.
A simple and reliable pretreatment protocol facilitates fluorescent in situ hybridisation on tissue microarrays of paraffin wax embedded tumour samples.
Molecular pathology : MP (2003-09-30)
S-F Chin, Y Daigo, H-E Huang, N G Iyer, G Callagy, T Kranjac, M Gonzalez, T Sangan, H Earl, C Caldas
ABSTRACT
To describe a robust pretreatment protocol for preparing paraffin wax embedded tissues on tissue microarrays for fluorescence in situ hybridisation (FISH). The newly developed pretreatment protocol described here was compared with the commonly used sodium thiocyanate based protocol and two different heating methods used in standard antigen unmasking protocols for immunohistochemistry (pressure cooking and microwaving in citrate acid buffer). Dewaxed tissue sections were incubated in 10mM citric acid buffer at 80 degrees C for 30 minutes to two hours, followed by a short pepsin digestion (1-5 mg/ml). Pretreated tissues were co-denatured with DNA probes at 80 degrees C for 10 minutes, followed by hybridisation at 37 degrees C for 48-72 hours. The three protocols using citrate acid buffer produced FISH signals with superior signal to noise ratios compared with sodium thiocyanate pretreatment. Most importantly, the best tissue attachment was achieved using the newly developed pretreatment protocol: on tissue microarrays less than 1% of cores were lost. To date, a total of 30 probes have been successfully hybridised on to breast tissue and multi-tissue microarrays. This pretreatment protocol is easy, reproducible, and facilitates FISH on tissue microarrays, with potential for widespread application in cancer research.
Sigma-Aldrich
Sodium thiocyanate solution, BioUltra, 8 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium thiocyanate, ACS reagent, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium thiocyanate, ≥99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium thiocyanate, reagent grade, 98-102% (titration)
Pricing and availability is not currently available.