MilliporeSigma
  • Interfacial conformation of dipalmitoylglycerol and dipalmitoylphosphatidylcholine in phospholipid bilayers.
Interfacial conformation of dipalmitoylglycerol and dipalmitoylphosphatidylcholine in phospholipid bilayers.
Biochemistry (1992-11-24)
S O Smith, I Kustanovich, S Bhamidipati, A Salmon, J A Hamilton
ABSTRACT
Diacylglycerols are minor constituents of membrane lipids, yet are essential in the activation and membrane association of protein kinase C. Solid-state 13C NMR experiments have been used to characterize the orientation of the glycerol backbone of dipalmitoylglycerol (DPG) and dipalmitoylphosphatidylcholine (DPPC) in egg phosphatidylcholine (PC) bilayers. The 13C NMR spectra of both DPG and DPPC specifically 13C-labeled at the sn-2 chain carbonyl exhibit a single narrow resonance (approximately 2 ppm) in liquid-crystalline egg PC bilayers. In contrast, specific 13C-labeling of both the sn-1 and sn-2 chain carbonyls results in an additional broad component (24-32 ppm) with an axially symmetric line shape. These data reveal that DPG has a distinct motionally-averaged structure in PC bilayers that is similar to that of DPPC and is not significantly affected by the absence of the large polar PC headgroup. The NMR line shapes are roughly consistent with the results of previous FTIR and NMR studies that indicate the sn-1 chain extends from the C1 carbon of the glycerol backbone into the hydrophobic interior of the bilayer, while the sn-2 chain first extends parallel to the bilayer surface and incorporates a bend at the ester linkage in order to keep the sn-1 and sn-2 chains parallel. However, the data suggest that the time-averaged orientation of the glycerol backbone is tilted from the bilayer normal, in contrast to the nearly parallel orientation observed in the crystal structures of phosphatidylcholines and phosphatidylethanolamines or the perpendicular orientation observed in the crystal structures of diacylglycerols.
Sigma-Aldrich
Dipalmitin, ≥99.0%