MilliporeSigma
  • A study of calcium ion-selective PVC membrane electrode based on neutral carrier N,n,n',n'-tetracyclo-3-oxapentanediamide (correction of oxapetanediamide).
A study of calcium ion-selective PVC membrane electrode based on neutral carrier N,n,n',n'-tetracyclo-3-oxapentanediamide (correction of oxapetanediamide).
Journal of Tongji Medical University = Tong ji yi ke da xue xue bao (1992-01-01)
Y L Ma, X H Rao, S M Zhong, S Ren, T X Yu, Q Zhen
ABSTRACT
A calcium ion-selective PVC membrane electrode based on neutral carrier n,n,n',n'-tetracyclohexyl-3-oxapetanediamide, using di-(2-ethylhexyl)phthalate as the plasticizer and potassium tetrakis (4-chlorophenyl) borate as the additive is reported in this paper. The ion selective membrane consists of 1 wt% of the Ca2+ selective ligand, 65 wt% of the plasticizer, 1 wt% of the additive and 33 wt% of poly (vinyl chloride) powder. The electrode has the linear response range of 2.0 x 10(-7)-10(-1) mol/L with the Nernstian slope of 28 mV/decade at 25 degrees C and the detection limit of 2.0 x 10(-8) mol/L. The response time of the calcium ion-selective electrode is as the concentration of calcium ion is rapidly shifted from 10(-5) to 10(-4) mol/L. The potential stability and reproducibility are good. The free calcium in blood serum was determined by the calcium ion-selective electrode with satisfactory results.
Product Description
Supelco
Calcium ionophore II, Selectophore, function tested