MilliporeSigma
  • Reveromycins, new inhibitors of eukaryotic cell growth. III. Structures of reveromycins A, B, C and D.
Reveromycins, new inhibitors of eukaryotic cell growth. III. Structures of reveromycins A, B, C and D.
The Journal of antibiotics (1992-09-01)
H Koshino, H Takahashi, H Osada, K Isono
ABSTRACT
Reveromycins A, B, C and D are new group inhibitors of the mitogenic activity of epidermal growth factor (EGF), produced by Streptomyces sp. Reveromycins are novel polyketide type antibiotics which have two terminal carboxylic groups, a spiroketal, a succinate and a varied side chain in the molecule. Determination of their structures by chemical and spectroscopic methods, in particular NMR studies, is described.
Sigma-Aldrich
Reveromycin A, solid