MilliporeSigma
  • Induction of CYP1A1 gene expression in H4-II-E rat hepatoma cells by benzo[e]pyrene.
Induction of CYP1A1 gene expression in H4-II-E rat hepatoma cells by benzo[e]pyrene.
Molecular carcinogenesis (1992-01-01)
W H Houser, A Raha, M Vickers
ABSTRACT
In the rat, expression of the CYP1A1 gene is closely associated with arylhydrocarbon hydroxylase (AHH) enzyme activity. AHH is an inducile enzyme activity known to play an important role in the bioactivation of polycyclic aromatic hydrocarbons (PAHs) to mutagenic and carcinogenic metabolites. PAH-induced expression of the CYP1A1 gene appears to be regulated by several trans-acting factors, including the Ah receptor and the 4S PAH-binding protein. In this study, we used the PAH isomers benzo[a]pyrene (BaP) and benzo[e]pyrene (BeP) to further evaluate the role of the 4S PAH-binding protein in induction of the CYP1A1 gene in H4-II-E rat hepatoma cells. Although BaP is believed to bind to both the Ah receptor and the 4S protein, BeP has been reported to bind exclusively to the 4S protein. The results of the study presented here indicate that BaP and BeP induce the expression of the CYP1A1 gene, as measured by ethoxyresorufin O-deethylase (EROD) activity, in a concentration-dependent manner. However, BaP is about 25 times as potent as BeP in inducing EROD activity in these cells. Slot-blot analysis of total RNA isolated from these cells indicated that BeP, BaP, and 3-methylcholanthrene increased the level of CYP1A1 mRNA expression. Sucrose-gradient analysis of BeP binding activity indicated that BeP bound with high affinity to the 4S PAH-binding protein, but not to the Ah receptor. These results suggest that the 4S protein may play a role in the PAH-induced expression of the CYP1A1 gene in rat H4-II-E cells.
Product Description
Supelco
Benzo[e]pyrene, analytical standard
Sigma-Aldrich
Benzo[e]pyrene, 98%
Benzo[e]pyrene, BCR®, certified reference material