MilliporeSigma
  • [Application of AOTF in spectral analysis. 3. Application of AOTF in atomic emission spectral analysis].
[Application of AOTF in spectral analysis. 3. Application of AOTF in atomic emission spectral analysis].
Guang pu xue yu guang pu fen xi = Guang pu (2003-08-27)
Ze-yong Chen, Rong-fei Peng, Zhan-xia Zhang
ABSTRACT
An atomic emission spectrometer based on acousto-optic tunable filter (AOTF) was self-constructed and was used to evaluate its practical use in atomic emission analysis. The AOTF used was of model TEAF5-0.36-0.52-S (Brimrose, USA) and the frequency of the direct digital RF synthesizer ranges from 100 MHz to 200 MHz. ICP and PMT were used as light source and detector respectively. The software, written in Visual C++ and running on the Windows 98 platform, is of an utility program system having two data banks and multiwindows. The wavelength calibration was performed with 14 emission lines of Ca, Y, Li, Eu, Sr and Ba using a tenth-order polynomial for line fitting method. The absolute error of the peak position was less than 0.1 nm, and the peak deviation was only 0.04 nm as the PMT varied from 337.5 V to 412.5 V. The scanning emission spectra and the calibration curves of Ba, Y, Eu, Sc and Sr are presented. Their average correlation coefficient was 0.9991 and their detection limits were in the range of 0.051 to 0.97 micrograms.mL-1 respectively. The detection limit can be improved under optimized operating conditions. However, the spectral resolution is only 2.1 nm at the wavelength of 488 nm. Evidently, this poor spectral resolution would restrict the application of AOTF in atomic emission spectral analysis, unless an enhancing techniques is integrated in it.
Sigma-Aldrich
Tellurium dioxide, ≥99%
Sigma-Aldrich
Tellurium dioxide, 99.995% trace metals basis
Sigma-Aldrich
Tellurium dioxide, ≥97.0%