MilliporeSigma
  • Efficient incorporation of positively charged 2', 3'-dideoxynucleoside-5'-triphosphates by DNA polymerases and their application in 'direct-load' DNA sequencing.
Efficient incorporation of positively charged 2', 3'-dideoxynucleoside-5'-triphosphates by DNA polymerases and their application in 'direct-load' DNA sequencing.
Nucleic acids research (2003-08-09)
Patrick J Finn, Matthew G Bull, Haiguang Xiao, Paula D Phillips, John R Nelson, Greg Grossmann, Satyam Nampalli, Bernard F McArdle, J Anthony Mamone, Parke K Flick, Carl W Fuller, Shiv Kumar
ABSTRACT
A series of charge-modified, dye-labeled 2', 3'-dideoxynucleoside-5'-triphosphates have been synthesized and evaluated as reagents for dye-terminator DNA sequencing. Unlike the commonly used dye-labeled terminators, these terminators possess a net positive charge and migrate in the opposite direction to dye-labeled Sanger fragments during electrophoresis. Post-sequencing reaction purification is not required to remove unreacted nucleotide or associated breakdown products prior to electrophoresis. Thus, DNA sequencing reaction mixtures can be loaded directly onto a separating medium such as a sequencing gel. The charge-modified nucleotides have also been shown to be more efficiently incorporated by a number of DNA polymerases than regular dye-labeled dideoxynucleotide terminators or indeed normal dideoxynucleoside-5'-triphosphates.
Sigma-Aldrich
Nε,Nε,Nε-Trimethyllysine hydrochloride, ≥97% (TLC)