MilliporeSigma
  • Lack of de-epoxidation of type B trichothecenes in incubates with human faeces.
Lack of de-epoxidation of type B trichothecenes in incubates with human faeces.
Food additives and contaminants (2003-07-26)
G Sundstøl Eriksen, H Pettersson
ABSTRACT
The ability of human gastrointestinal organisms to transform the trichothecenes 3-acetyldeoxynivalenol and nivalenol was investigated. Samples of human faeces were incubated under anaerobic conditions for 48 h with the toxins. They were then extracted and analysed for trichothecenes and metabolites. 3-acetyldeoxynivalenol was metabolized to deoxynivalenol during the incubation period. In contrast to what has been reported for other species such as rats, mice and pigs, no de-epoxidated metabolites were detected in the faecal incubates. The toxicological significance of the difference in the intestinal ability to transform trichothecenes between species is unknown.
Product Description
Supelco
3-Acetyldeoxynivalenol solution, ~100 μg/mL in acetonitrile, analytical standard
Sigma-Aldrich
3-Acetyldeoxynivalenol, from Fusarium roseum