MilliporeSigma
  • Interaction between isopoly-tungstic acid and berberine hydrochloride by resonance Rayleigh scattering spectrum.
Interaction between isopoly-tungstic acid and berberine hydrochloride by resonance Rayleigh scattering spectrum.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2003-07-26)
Shaopu Liu, Ling Kong
ABSTRACT
In a weak acid medium, when an isopoly-tungstic acid reacts with berberine to form an ion-association complex, strong resonance Rayleigh scattering (RRS) appears and obvious frequency-doubling scattering (FDS) as well as second-order scattering (SOS) come into being. The scattering peaks are located at 288 and 369 nm for RRS, at 290, 370 and 390 nm for FDS, and at 480, 540 and 740 nm for SOS. Under optimum conditions, all three scattering intensities are directly proportional to the concentration of berberine and can be applied to the determination of berberine. The sensitivity of the RRS method is the highest; the detection limit (3sigma) for berberine hydrochloride is 7.6 ng/ml. The method also has good selectivity. It has been applied to the determination of berberine in pharmaceuticals and goldthread extracts with satisfactory results. In the paper, studies of the existing species of isopoly-acid and the component of the ion-association complex are reported and the reasons of enhancement of RRS are discussed.
Sigma-Aldrich
Tungstic acid, 99%
Sigma-Aldrich
Tungstic acid, ≥98.0% (calcined substance, T), powder