MilliporeSigma
Facile preparation of DNA-tagged carbohydrates.
Bioorganic & medicinal chemistry letters (2003-07-23)
Shinsuke Sando, Kazuki Matsui, Yusuke Niinomi, Nobuhiko Sato, Yasuhiro Aoyama
ABSTRACT
We report here that unprotected carbohydrates (maltose, lactose, cellobiose, and maltoheptaose) can be attached to the aminoalkylated oligonucleotides under mild reductive-amination conditions (aqueous borate buffer, pH 8.0, NaBH(3)CN, 60 degrees C) without notable side reactions. Quadruplex-forming G-rich oligonucleotide, 5'-aminoalkyl d(TGGGGT), is glycosylated with maltoheptaose to afford a novel DNA-assisted tetrasaccharide cluster motif.
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%