MilliporeSigma
  • A practical and convenient synthesis of hotrienol, an excellent fruity smelling compound.
A practical and convenient synthesis of hotrienol, an excellent fruity smelling compound.
Journal of agricultural and food chemistry (2003-06-26)
Yoshifumi Yuasa, Yasushi Kato
ABSTRACT
The practical and convenient synthesis of hotrienol, which is an excellent fruity smelling compound, has been performed by the ene-type chlorination of linalyl acetate and then dehydrochlorinated by lithium bromide and lithium carbonate in DMF, followed by hydrolysis in three steps with an overall yield of 55%.
Sigma-Aldrich
Linalyl acetate, natural, ≥96%, FG
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O
Sigma-Aldrich
3,7-Dimethyl-1,6-octadien-3-yl acetate, 97%
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis