MilliporeSigma
  • Differences in alpha 2u-globulins increased in male rat kidneys following treatment with several alpha 2u-globulin accumulating agents: cystein protease(s) play(s) an important role in production of kidney-type-alpha 2u-globulin.
Differences in alpha 2u-globulins increased in male rat kidneys following treatment with several alpha 2u-globulin accumulating agents: cystein protease(s) play(s) an important role in production of kidney-type-alpha 2u-globulin.
Toxicology (1992-11-30)
K Saito, H Kaneko, N Isobe, I Nakatsuka, A Yoshitake, H Yamada
ABSTRACT
Effects of alpha 2u-globulin accumulating agents on alpha 2u-globulins in rat kidneys were examined by sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) and immunoblotting analysis. Treatment of male animals with decalin (150 mg/kg), 2,2,4-trimethylpentane (50 mg/kg), isophorone (150 mg/kg), d-limonene (150 mg/kg) or 1,4-dichlorobenzene (150 mg/kg) by gavage for 14 consecutive days in each case resulted in a marked intensification of a protein band corresponding to kidney-type-alpha 2u-globulin, with a molecular mass calculated to be approximately 16 kDa. However, intraperitoneal treatment with leupeptin and E-64 (two times 0.07 mmol/kg, for each), well known cystein protease inhibitors, while only slightly increasing this kidney-type-alpha 2u-globulin band, caused the intensification of a approximately 19-kDa molecular mass protein band which was revealed to be a native-type-alpha 2u-globulin by SDS-PAGE and immunoblotting. These results indicated that at least two types of alpha 2u-globulin can be increased in male rat kidney by chemical treatment. Moreover, cystein protease(s) appear(s) to play an important role in the degradation of alpha 2u-globulin and particularly in the conversion of native-type-alpha 2u-globulin to kidney-type-alpha 2u-globulin in rat kidneys.
Sigma-Aldrich
Isophorone, 97%
Sigma-Aldrich
Isophorone, ≥97%, FG