MilliporeSigma
Fragrance material review on linalyl acetate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2003-06-14)
C S Letizia, J Cocchiara, J Lalko, A M Api
ABSTRACT
A toxicologic and dermatologic review of linalyl acetate when used as a fragrance ingredient is presented.
Sigma-Aldrich
Linalyl acetate, natural, ≥96%, FG
Supelco
Linalyl acetate, analytical standard
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG
Sigma-Aldrich
3,7-Dimethyl-1,6-octadien-3-yl acetate, 97%