MilliporeSigma
  • Antagonism of 3,4-methylenedioxymethamphetamine-induced neurotoxicity in rat brain by 1-piperonylpiperazine.
Antagonism of 3,4-methylenedioxymethamphetamine-induced neurotoxicity in rat brain by 1-piperonylpiperazine.
European journal of pharmacology (1992-09-01)
K Hashimoto, H Maeda, T Goromaru
ABSTRACT
The effects of 1-piperonylpiperazine and N,alpha-dimethylpiperonylamine, which are weak inhibitors for [3H]5-hydroxytryptamine (5-HT) uptake, on 3,4-methylenedioxymethamphetamine (MDMA)-induced neurotoxicity were examined. The reductions of serotonergic parameters in the rat cerebral cortex produced by multiple administration of MDMA (10 mg/kg) were attenuated significantly by coadministration of 6-nitroquipazine (10 mg/kg), paroxetine (10 mg/kg) or 1-piperonylpiperazine (20 mg/kg), but not by N,alpha-dimethylpiperonylamine (20 mg/kg). The present data suggest that 1-piperonylpiperazine might inhibit the MDMA-induced neurotoxicity by effect(s) other than 5-HT uptake inhibition.
Sigma-Aldrich
1-Piperonylpiperazine, 97%