MilliporeSigma
Ycf1p-dependent Hg(II) detoxification in Saccharomyces cerevisiae.
European journal of biochemistry (2003-05-21)
Olivier Gueldry, Myriam Lazard, Florence Delort, Marc Dauplais, Ioana Grigoras, Sylvain Blanquet, Pierre Plateau
ABSTRACT
In Saccharomyces cerevisiae, disruption of the YCF1 gene increases the sensitivity of cell growth to mercury. Transformation of the resulting ycf1 null mutant with a plasmid harbouring YCF1 under the control of the GAL promoter largely restores the wild-type resistance to the metal ion. The protective effect of Ycf1p against the toxicity of mercury is especially pronounced when yeast cells are grown in rich medium or in minimal medium supplemented with glutathione. Secretory vesicles from S. cerevisiae cells overproducing Ycf1p are shown to exhibit ATP-dependent transport of bis(glutathionato)mercury. Moreover, using beta-galactosidase as a reporter protein, a relationship between mercury addition and the activity of the YCF1 promoter can be shown. Altogether, these observations indicate a defence mechanism involving an induction of the expression of Ycf1p and transport by this protein of mercury-glutathione adducts into the vacuole. Finally, possible coparticipation in mercury tolerance of other ABC proteins sharing close homology with Ycf1p was investigated. Gene disruption experiments enable us to conclude that neither Bpt1p, Yor1p, Ybt1p nor YHL035p plays a major role in the detoxification of mercury.
Sigma-Aldrich
Mercury(II) bromide, ACS reagent
Sigma-Aldrich
Mercury(II) bromide, puriss. p.a., ACS reagent, ≥99.0% (precipitation titration)