MilliporeSigma
  • High performance carbon-supported catalysts for fuel cells via phosphonation.
High performance carbon-supported catalysts for fuel cells via phosphonation.
Chemical communications (Cambridge, England) (2003-05-13)
Zhiqiang Xu, Zhigang Qi, Arthur Kaufman
ABSTRACT
Carbon-supported catalysts were phosphonated using 2-aminoethylphosphonic acid, and the resulting catalysts with largely enhanced proton conductivity performed substantially better than the untreated counterparts in proton-exchange membrane fuel cells.
Sigma-Aldrich
2-Aminoethylphosphonic acid, 99%