MilliporeSigma
  • IR spectral and structural changes caused by the conversion of 3-hydroxybenzaldehyde into the oxyanion.
IR spectral and structural changes caused by the conversion of 3-hydroxybenzaldehyde into the oxyanion.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-04-26)
B A Stamboliyska, E A Velcheva, I G Binev
ABSTRACT
The spectral and structural changes, caused by the conversion of the 3-hydroxybenzaldehyde molecule into the corresponding oxyanion have been studied by IR spectra, and MP2 and DFT force field calculations. The conversion causes a 13 cm(-1) decrease in the frequency of the carbonyl stretching band nu(Cz=O), a 1.3-fold increase in its integrated intensity, strong intensity increases (2.1-5.3-fold) of the aromatic skeletal nu8 and nu19 as well as formyl nu(CH) bands. According to the calculations the oxyanionic charge is delocalized over aldehyde group (0.37 e-), phenylene ring (0.12 e-) and oxyanionic center (0.52 e-). The conversion into the oxyanion leads to geometry changes in the whole species, but it remains planar.
Sigma-Aldrich
3-Hydroxybenzaldehyde, ≥99%