MilliporeSigma
  • Vibrational study of the secondary thioamide function, the intramolecular resonance assisted and intermolecular hydrogen bonding in NN'-hydroxyalkyl dithiooxamides.
Vibrational study of the secondary thioamide function, the intramolecular resonance assisted and intermolecular hydrogen bonding in NN'-hydroxyalkyl dithiooxamides.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2003-03-28)
H O Desseyn, N Blaton, B Slootmaekers, S P Perlepes
ABSTRACT
The article describes the vibrational characterization of the secondary thioamide formation and especially a spectroscopical interesting study of different types of hydrogen bonding in NN'-dihydroxyalkyldithiooxamides.
Sigma-Aldrich
Dithiooxamide, 97%