MilliporeSigma
  • Modelling the effect of oil/fat content in food systems on flavour absorption by LLDPE.
Modelling the effect of oil/fat content in food systems on flavour absorption by LLDPE.
Food additives and contaminants (2003-03-08)
M Dekker, R W G van Willige, J P H Linssen, A G J Voragen
ABSTRACT
One of the phenomena in food-packaging interactions is flavour absorption. Absorption of flavour compounds from food products into food-packaging materials can result in loss of flavour compounds or an unbalance in the flavour profile changing a product's quality. The food matrix influences the amounts of absorbed flavour compounds; the presence of oil or fat especially determines the ability to absorb flavour compounds from the food to the package. On the other hand, the polarity of the flavour compound itself is a characteristic that also influences the level of absorption into synthetic polymers. A model based on the effect of the polarity (logP) of flavour compounds and on their partitioning coefficients between the food (matrix) and the packaging material is described. The model can be used for predicting absorption of flavour compounds from foods into LLDPE. However, an attempt to apply the proposed model on real foods shows serious limitations of the model for (very) low fat products. Predictive values deviate from the measured values, probably due to other interaction phenomena, e.g. with proteins. Predictive and measured values from a product with a substantial amount of fat match much better, suggesting that the model is valid for products having a substantial amount of (free) fat.
Sigma-Aldrich
Decanal, FCC, FG
Sigma-Aldrich
Decanal, ≥98% (GC), liquid
Sigma-Aldrich
Decanal, natural, ≥97%, FG