MilliporeSigma
  • Crystallization of beta-galactosidase does not reduce the range of activity of individual molecules.
Crystallization of beta-galactosidase does not reduce the range of activity of individual molecules.
Biochemistry (2003-02-13)
Glen K Shoemaker, Douglas H Juers, Jennifer M L Coombs, Brian W Matthews, Douglas B Craig
ABSTRACT
By use of a capillary electrophoresis-based procedure, it is possible to measure the activity of individual molecules of beta-galactosidase. Molecules from the crystallized enzyme as well as the original enzyme preparation used to grow the crystals both displayed a range of activity of 20-fold or greater. beta-Galactosidase molecules obtained from two different crystals had indistinguishable activity distributions of 31,600 +/- 1100 and 31,800 +/- 1100 reactions min(-1) (enzyme molecule)(-1). This activity was found to be significantly different from that of the enzyme used to grow the crystals, which showed an activity distribution of 38,500 +/- 900 reactions min(-1) (enzyme molecule)(-1).
Sigma-Aldrich
Resorufin β-D-galactopyranoside, ~95%