MilliporeSigma
  • The effect of ivomec super on dogs experimentally infested with Sarcoptes scabiei isolated from a patient.
The effect of ivomec super on dogs experimentally infested with Sarcoptes scabiei isolated from a patient.
Journal of the Egyptian Society of Parasitology (2003-02-01)
T A Morsy, M Y Abdel-Mawla, A T Morsy
ABSTRACT
Scabies infestation is a community health problem worldwide. This is particularly true in overcrowded and unhygienic areas. Seven dogs were experimentally infested with Sarcoptes scabiei freshly recovered from a patient. The dogs after infestation were treated with a mixture of ivermectin and clorsulon (Ivomec Super). The dose given to a single dog was 1 ml/50 kg body weight. This is equivalent to 200 mcg ivermectin and 2 mg clorsulon per Kg body weight. Five of the dogs (71.43%) were completely cured with 1 ml/50 Kg body weight. The remaining two dogs needed each, another injection of the same dose two weeks later. So, ivomec super is effective against the human strain of Sarcoptes scabiei in experementally infested dogs. A general discussion was given.
Product Description
Supelco
Clorsulon, VETRANAL®, analytical standard