MilliporeSigma
  • Cytotoxic effects of the polyamine oxidase inactivator MDL 72527 to two human colon carcinoma cell lines SW480 and SW620.
Cytotoxic effects of the polyamine oxidase inactivator MDL 72527 to two human colon carcinoma cell lines SW480 and SW620.
Cell biology and toxicology (2002-12-18)
B Duranton, V Holl, Y Schneider, S Carnesecchi, F Gossé, F Raul, N Seiler
ABSTRACT
N1,N4-bis(2,3-butadienyl)-1,4-butanediamine (MDL 72527) was considered to be a selective inactivator of FAD-dependent tissue polyamine oxidase. Recently MDL 72527 was reported to induce apoptosis in transformed hematopoietic cells through lysosomotropic effects. Since it is the only useful inhibitor of polyamine oxidase available at present, the re-evaluation of its properties seemed important. Human colon carcinoma-derived SW480 cells and their lymph node metastatic derivatives (SW620) were chosen for our study because they differ in various aspects of polyamine metabolism but have similar polyamine oxidase activities. MDL 72527 inhibited cell growth in a concentration-dependent manner, depleted intracellular polyamine pools, and caused the accumulation of N1-acetyl derivatives of spermidine and spermine. SW620 cells were more sensitive to the drug than were SW480 cells. At 150 micromol/L MDL 72527, SW620 cells accumulated in S-phase of the cell cycle, showed decreased polyamine transport rate, and showed no increase of polyamine N1-acetyltransferase activity. In contrast, SW480 cells were not arrested in a particular phase of the cell cycle, showed enhanced polyamine uptake, and showed a mild induction of acetyltransferase. The results suggest that MDL 72527 retains its value as a selective tool in short-term experiments only at concentrations not exceeding those necessary for the inactivation of polyamine oxidase. At concentrations above 50 micromol/L and at exposure times longer than 24 h, it may derange cell functions nonspecifically, and thus blur the results of studies intended to elucidate polyamine oxidase functions.
Sigma-Aldrich
MDL 72527, ≥98% (HPLC)