MilliporeSigma
  • [Construction and identification for cell strain of anti-human seminal plasma phospholipase A2 monoclonal antibody].
[Construction and identification for cell strain of anti-human seminal plasma phospholipase A2 monoclonal antibody].
Zhonghua nan ke xue = National journal of andrology (2002-12-14)
Shu-Kui Wang, Yu-Feng Huang, Xing-Yi Xia, Bao-Tong Li, Zi-Zheng Wang, Meng-Liu Liu
ABSTRACT
To establish and evaluate the anti-human seminal plasma phospholipase A2 (PLA2) monoclonal antibody (McAb). After having been separated and purified from human seminal plasma by PEG precipitation, Sephacryl S-300 column chromatography, DEAE-Sephadex A-25 column chromatography and HA column chromatography, PLA2 was regarded as an antigen to immune BALB/C mouse to produce anti-human seminal plasma PLA2 McAb. The PLA2 McAb sensitivity and specificity were performed by ELISA technique and Western-blot analysis, respectively. The molecular weight of PLA2 depurated with 245 fold purification from human seminal plasma was about 34,900, while the sensitivity and typing of its McAb were 1:5(6)-1:5(8) and IgM (kappa) with a satisfied Western-Blot results. The PLA2, which had not been reported in international and domestic papers, may be a new type of PLA2. The establish of its McAb will provide significant tools for the research of the relationship between PLA2 in human seminal plasma and male fertility.
Sigma-Aldrich
DEAE–Sephadex® A-25 chloride form