MilliporeSigma
  • [FeII(bpy)3]2+/TiO2-codoped zeolites: synthesis, characterization, and first application in photocatalysis.
[FeII(bpy)3]2+/TiO2-codoped zeolites: synthesis, characterization, and first application in photocatalysis.
Chemphyschem : a European journal of chemical physics and physical chemistry (2002-12-06)
Stefan H Bossmann, Nabil Shahin, Hieu Le Thanh, Anna Bonfill, Michael Wörner, André M Braun
ABSTRACT
TiO2-codoped [FeII(bpy)3]2+/zeolite Y systems are revealed to be efficient photocatalysts for photochemically enhanced Fenton processes. Preparation, characterization, and tests using 2,4-xylidine as a model pollutant are described, and a first mechanistic hypothesis explaining the rather high photochemical reactivity of the catalyst system is given.
Sigma-Aldrich
2,4-Dimethylaniline, ≥99%