MilliporeSigma
[Initial stages of trisporic acid synthesis in Blakeslea trispora].
Prikladnaia biokhimiia i mikrobiologiia (2002-11-27)
N N Gessler, A V Sokolov, T A Belozerskaia
ABSTRACT
Biotransformation of beta-carotene with enzyme preparations isolated from the mycelium of Blakeslea trispora resulted in the formation of its hydroxylated metabolite and apocarotenals, products of oxidative degradation of this compound. By spectral, chromatographic, and chemical properties, the beta-carotene derivative was identified as 4-hydroxy-beta-carotene (isocryptoxanthine). One of the products of oxidative degradation of beta-carotene, beta-apo-13-carotenone, underwent modification in the presence of enzyme preparations from Blakeslea trispora with the formation of trisporic acid precursors. It should be emphasized that beta-apo-13-carotenone transformation proceeded more rapidly than beta-carotene oxidation by carbon in the 4-position. Our findings suggest that under conditions of oxidative stress, oxidative degradation of beta-carotene into beta-apo-13-carotenone leads to the formation of considerable amounts of trisporic acids.
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ≥96.0% (UV)
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ~20% apocarotenal basis (UV-vis), suspension (oily)