MilliporeSigma
  • Products of the determination of the iodine value with iodine monobromide.
Products of the determination of the iodine value with iodine monobromide.
Archiv der Pharmazie (2002-11-26)
Peter Imming, Oliver Germershaus
ABSTRACT
The iodine value (iodine number) is an important analytical characteristic of fats and oils. Leading pharmacopeias determine it using iodine monobromide (Hanus method). We used methyl oleate as a simple analog of unsaturated triacylglycerols to identify the products. After performing the reaction in deuterated solvents under pharmacopeial conditions, NMR spectroscopy revealed the presence of the 9, 10-diiodo, 9, 10-dibromo, and 9, 10-bromoiodo adducts, leaving no educt olefin. The prescribed subsequent addition of potassium iodide led to the formation of methyl 9, 10-diiodo and bromoiodo stearate in equal amounts.
Sigma-Aldrich
Iodine monobromide, 98%