MilliporeSigma
Nephrotoxicity of chlorofluoroacetic acid in rats.
Toxicological sciences : an official journal of the Society of Toxicology (2002-11-21)
Hoffman B M Lantum, Raymond B Baggs, Daria M Krenitsky, M W Anders
ABSTRACT
Dichloroacetic acid (DCA), chlorofluoroacetic acid (CFA), and difluoroacetic acid (DFA) are inhibitors of pyruvate dehydrogenase kinase. DCA is used for the clinical management of congenital lactic acidosis. Glutathione transferase zeta (GSTZ1-1) catalyzes the biotransformation of DCA and CFA, and DCA is a mechanism-based inactivator of GSTZ1-1. In rodents, DCA causes multiorgan toxicities and is hepatocarcinogenic. The toxic effects of CFA, which is an excellent substrate but a poor inactivator of GSTZ1-1, have not been investigated. The objective of this study was to investigate the nephrotoxicity of CFA. Rats given a single dose of 1.5 mmol/kg CFA became anuric and died within 24 h. Urinalysis and light microscopic analysis showed that rats given 0.6-1.2 mmol/kg CFA developed polyuria, glycosuria, and renal proximal tubular damage. Electron microscopic analysis indicated a role for apoptosis in CFA-induced cell death. The nephrotoxicity of CFA was associated with a dose-dependent increase in inorganic fluoride excretion. Treatment of rats with DCA for 5 days to inactivate GSTZ1-1 failed to prevent metabolism of CFA to fluoride and did not block CFA-induced renal damage. A role for GSTZ1-1-catalyzed release of fluoride from CFA is proposed but a role for other enzymes cannot be excluded. DFA, which is not metabolized to fluoride by GSTZ1-1, was given to rats as a control and was also nephrotoxic: rats given 1.2 mmol DFA/kg/day for 5 days had normal urine volumes but showed proximal and distal tubular damage; fluoride excretion was not elevated. The mechanism of DFA-induced nephrotoxicity is not known but appears to differ from that of CFA.
Sigma-Aldrich
Difluoroacetic acid, 98%