MilliporeSigma
  • Acid hydrolysis of silk fibroins and determination of the enrichment of isotopically labeled amino acids using precolumn derivatization and high-performance liquid chromatography-electrospray ionization-mass spectrometry.
Acid hydrolysis of silk fibroins and determination of the enrichment of isotopically labeled amino acids using precolumn derivatization and high-performance liquid chromatography-electrospray ionization-mass spectrometry.
Analytical biochemistry (2002-11-21)
Sonja Hess, Jacco van Beek, Lewis K Pannell
ABSTRACT
Silk fibroins from moth larvae and spiders are composed of highly repetitive Ala- and Gly-rich blocks that determine their structure, properties, and function. To investigate the metabolic integration of isotopically labeled amino acids in the excreted silk, the enrichment of ingested tracers was determined after acid hydrolysis of the fibroins. Thus, spiders and moth larvae were fed with stable isotope tracers such as [1-13C]Ala or [1-13C]Gly and silked. After hydrolysis of the silk proteins, the corresponding amino acids were derivatized with Nalpha-(2,4-dinitro-5-fluorophenyl)-L-alaninamide (Marfey's reagent) and separated by liquid chromatography. The isotopical enrichment of the amino acids was determined by online electrospray mass spectrometry and calculated by newly developed software. Depending on the feeding protocol, enrichments of up to 58% in Gly and 31% in Ala were found in the investigated silks. The highly enriched silk fibroins are suitable for further structural investigation such as solid-state nuclear magnetic resonance.
Sigma-Aldrich
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, powder
Supelco
Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, LiChropur, ≥99.0%