MilliporeSigma
Homocysteine promotes the LDL oxidase activity of ceruloplasmin.
FEBS letters (2002-11-19)
M Exner, M Hermann, R Hofbauer, B Hartmann, S Kapiotis, B Gmeiner
ABSTRACT
Ceruloplasmin (CP) oxidises low density lipoprotein (LDL). The oxidising potential depends on the formation of Cu(+)-CP which is redox-cycled during oxidation. Homocysteine (HCY) reduces free Cu(2+), potentiating its cell-damaging property. We show that HCY enhanced LDL oxidation by CP, but did not activate the LDL oxidising potential of Cu(2+)-diamine oxidase. Selective removal of the redox-active Cu(2+) abolished the LDL oxidase activity of CP. However, HCY partially restored the LDL oxidase activity of redox-copper depleted CP, indicating that the remaining six copper atoms in CP may also be involved in the process. Spectroscopic and oxidation inhibition studies using the Cu(+)-reagent bathocuproine revealed that HCY induced Cu(+)-CP formation, thus promoting its LDL oxidase activity.
Sigma-Aldrich
Bathocuproinedisulfonic acid disodium salt, for spectrophotometric det. of Cu, Fe