MilliporeSigma
  • Modification of the upper rim of homooxacalix[3]arenes and complexation between a nitrohomooxacalix[3]arene derivative and n-hexylamine.
Modification of the upper rim of homooxacalix[3]arenes and complexation between a nitrohomooxacalix[3]arene derivative and n-hexylamine.
The Journal of organic chemistry (2002-11-09)
Kazunori Tsubaki, Tadamune Otsubo, Tatsuya Morimoto, Hiromi Maruoka, Mika Furukawa, Yashima Momose, Muhong Shang, Kaoru Fuji
ABSTRACT
Several functional groups were introduced on the upper rim of (lower rim free) homooxacalix[3]arene for the first time. The swinging nitrohomooxacalix[3]arene host 1 was fixed in the cone conformation by complexation with n-hexylamine.
Sigma-Aldrich
Hexylamine, 99%