MilliporeSigma
High resolution emission spectra of TL materials.
Radiation protection dosimetry (2002-10-18)
T Vilaithong, S Wanwilairat, M Rhodes, W Hoffmann, T Messarius
ABSTRACT
High resolution emission spectra of several TL materials, that are commonly used in dosimetry, were measured using a low cost fibre optic spectrometer containing a 2048 pixel CCD array. The spectra were taken from 177 to 890 nm with a wavelength resolution of 1.3 nm. This allowed for line width discrimination and the resolution of transitions that have not been seen so far. For rare earth doped materials like CaF2:Tm (TLD-300) and CaF2:Dy (TLD-200) the spectral lines were compared to the energy levels measured by Dieke and Crosswhite leading to the identification of most of the transition lines.
Sigma-Aldrich
Lithium tetraborate, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium tetraborate, ≥99.995% trace metals basis
Sigma-Aldrich
Lithium metaborate, 99.9% trace metals basis
Sigma-Aldrich
Lithium metaborate, 99.995% trace metals basis
Sigma-Aldrich
Lithium metaborate, ACS reagent, ≥98.0%