MilliporeSigma
  • Enzymatic formation of an unnatural C(6)-C(5) aromatic polyketide by plant type III polyketide synthases.
Enzymatic formation of an unnatural C(6)-C(5) aromatic polyketide by plant type III polyketide synthases.
Organic letters (2002-10-12)
Ikuro Abe, Yusuke Takahashi, Hiroshi Noguchi
ABSTRACT
[reaction: see text] Substrate specificities of plant polyketide synthases (PKSs) were investigated using analogues of malonyl-CoA, the extension unit of the polyketide chain elongation reactions. When incubated with methylmalonyl-CoA and 4-coumaroyl-CoA, plant PKSs (chalcone synthase from Scutellaria baicalensis, stilbene synthase from Arachis hypogaea, and benzalacetone synthase from Rheum palmatum) afforded an unnatural C(6)-C(5) aromatic polyketide, 1-(4-hydroxyphenyl)pent-1-en-3-one, formed by one-step decarboxylative condensation of the two substrates. In contrast, succinyl-CoA was not accepted as a substrate by the enzymes.
Sigma-Aldrich
Benzylideneacetone, ≥98%, FG
Sigma-Aldrich
4-Phenyl-3-buten-2-one, 99%
Sigma-Aldrich
trans-4-Phenyl-3-buten-2-one, ≥99%