MilliporeSigma
  • Attachment of neutrals during tandem mass spectrometry of sulfonic acid dyes and intermediates in an ion trap.
Attachment of neutrals during tandem mass spectrometry of sulfonic acid dyes and intermediates in an ion trap.
Journal of mass spectrometry : JMS (2002-10-11)
Adrian Weisz, Denis Andrzejewski, Henry M Fales, Asher Mandelbaum
ABSTRACT
Several positional isomers of 2-(2-quinolinyl)-1H-indene-1,3(2H)-dione mono- and disulfonic acids prepared as reference materials for development of analytical methods involved in FDA certification of D&C Yellow No. 10 (Quinoline Yellow) were found consistently to show [MH + 14](+) ions when their electrospray- or atmospheric pressure chemical ionization-prepared MH(+) ions were subjected to collisional activation. The source of these ions was found to be the methanol used as solvent in these procedures which combined with their [MH - H(2)O](+) ions under chemical ionization conditions. The reaction was found to be sensitive to their isomeric and chemical structures and other examples of this process are reviewed.
Sigma-Aldrich
Quinoline Yellow, Mixture of the mono- and disulfonic acids of Quinoline Yellow
Sigma-Aldrich
Quinoline Yellow, mixture of mono- and disulfonic acid sodium salt