MilliporeSigma
  • Ion-molecule interactions in solutions of lithium perchlorate in propylene carbonate + diethyl carbonate mixtures: an IR and molecular orbital study.
Ion-molecule interactions in solutions of lithium perchlorate in propylene carbonate + diethyl carbonate mixtures: an IR and molecular orbital study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2002-09-06)
Jianji Wang, Yanping Wu, Xiaopeng Xuan, Hanqing Wang
ABSTRACT
FTIR spectra have been recorded and analyzed for solutions of lithium perchlorate in propylene carbonate (PC), diethyl carbonate (DEC), and PC + DEC mixtures. It has been shown that the carbonyl stretch bands for PC and DEC are very sensitive to the interaction between Li+ and the solvent molecules. They split with addition of LiClO4, indicating a strong interaction of Li+ with PC and DEC through the oxygen group of PC and both oxygen and ether oxygen atoms of DEC. In conjunction with molecular orbital calculation, the optimized geometries of solvation are given. In addition, solvent separated ion pairs and contact ion pairs were observed in LiClO4/DEC solutions, and no preferential solvation of Li+ in LiClO4/PC + DEC solutions were detected.
Sigma-Aldrich
Diethyl carbonate, 99%
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Diethyl carbonate, anhydrous, ≥99%
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, ACS reagent, ≥95.0%
Sigma-Aldrich
Lithium perchlorate, purum p.a., ≥98.0% (calc. based on dry substance, T), powder