MilliporeSigma
  • LC-MS determination of urinary toluenediamine in workers exposed to toluenediisocyanate.
LC-MS determination of urinary toluenediamine in workers exposed to toluenediisocyanate.
Toxicology letters (2002-08-23)
Tadashi Sakai, Y Morita, Y Kim, Y X Tao
ABSTRACT
To improve the biological monitoring method for 2,6- and 2,4-toluenediisocyanate (TDI) exposure, we developed a simple and rapid method for analysis of the corresponding urinary metabolites, 2,6- and 2,4-toluenediamine (TDA) using liquid chromatograph-mass spectrometry (LC-MS). One ml of urine was hydrolyzed at 100 degrees C for 1.5 h with H(2)SO(4). Alkalinized hydrolysate was extracted with dichloromethane (DCM) and analyzed by atmospheric pressure chemical ionization (APCI) LC-MS, in positive-ion mode. The mass spectra of TDA isomers showed the protonated molecule [M+H](+), at m/z 123 as the base peak. Calibration curves of 2,6-TDA were linear up to 400 microg/l. TDA isomers in urine of exposed workers as determined by LC-MS correlated well with those obtained by gas chromatography-mass spectrometry. 2,6- and 2,4-TDA were not detected in non-exposed subjects, whereas exposed workers showed urinary levels up to 250 and 63 microg/l, respectively.
Sigma-Aldrich
Tolylene-2,6-diisocyanate, 97%
Sigma-Aldrich
2,4-Diaminotoluene, 98%
Supelco
2,6-TDI, analytical standard
Supelco
2-Methyl-m-phenylenediamine, analytical standard
Sigma-Aldrich
2,6-Diaminotoluene, 97%