MilliporeSigma
  • Identification of potent odorants in Chinese jasmine green tea scented with flowers of Jasminum sambac.
Identification of potent odorants in Chinese jasmine green tea scented with flowers of Jasminum sambac.
Journal of agricultural and food chemistry (2002-08-09)
Yuriko Ito, Akio Sugimoto, Takami Kakuda, Kikue Kubota
ABSTRACT
The odorants in Chinese jasmine green tea scented with jasmine flowers (Jasminum sambac) were separated from the infusion by adsorption to Porapak Q resin. Among the 66 compounds identified by GC and GC/MS, linalool (floral), methyl anthranilate (grape-like), 4-hexanolide (sweet), 4-nonanolide (sweet), (E)-2-hexenyl hexanoate (green), and 4-hydroxy-2,5-dimethyl-3(2H)-furanone (sweet) were extracted as potent odorants by an aroma extract dilution analysis and sensory analysis. The enantiomeric ratios of linalool in jasmine tea and Jasminum sambac were determined by a chiral analysis for the first time in this study: 81.6% ee and 100% ee for the (R)-(-)-configuration, respectively. The jasmine tea flavor could be closely duplicated by a model mixture containing these six compounds on the basis of a sensory analysis. The omission of methyl anthranilate and the replacement of (R)-(-)-linalool by (S)-(+)-linalool led to great changes in the odor of the model. These two compounds were determined to be the key odorants of the jasmine tea flavor.
Sigma-Aldrich
Methyl 2-aminobenzoate, ReagentPlus®, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Methyl anthranilate, ≥98%, FCC, FG
Pricing and availability is not currently available.