MilliporeSigma
  • Quantum chemical- and 3-D-QSAR (CoMFA) studies of benzalacetones and 1,1,1-trifluoro-4-phenyl-3-buten-2-ones.
Quantum chemical- and 3-D-QSAR (CoMFA) studies of benzalacetones and 1,1,1-trifluoro-4-phenyl-3-buten-2-ones.
Bioorganic & medicinal chemistry letters (2002-08-06)
Chisako Yamagami, Noriko Motohashi, Miki Akamatsu
ABSTRACT
The inhibitory effect (IC(50)) of the title compounds on UV-induced mutagenesis in Escherichia coli WP2uvrA was analyzed quantitatively by using various quantum chemical descriptors and also by the CoMFA method: both approaches provided results of similar quality. The activity was shown to be increased by electron-withdrawing substituents and also by hydrogen-bonding between 2-hydroxy group and the bio-component.
Sigma-Aldrich
Benzylideneacetone, ≥98%, FG
Sigma-Aldrich
trans-4-Phenyl-3-buten-2-one, ≥99%
Sigma-Aldrich
4-Phenyl-3-buten-2-one, 99%