MilliporeSigma
  • The activation of neuronal nitric-oxide synthase by various divalent cations.
The activation of neuronal nitric-oxide synthase by various divalent cations.
The Journal of pharmacology and experimental therapeutics (2002-07-20)
John Weaver, Supatra Porasuphatana, Pei Tsai, Guan-Liang Cao, Theodore A Budzichowski, Linda J Roman, Gerald M Rosen
ABSTRACT
Nitric-oxide synthase (NOS; EC 1.14.13.39) catalyzes the oxidation of L-arginine to nitric oxide (NO(.)) and L-citrulline via the intermediate N(omega)-hydroxy-L-arginine. Of the three distinct isoforms of NOS that have been characterized, the constitutive neuronal NOS (NOS I) generates NO(.) associated with long-term potentiation (LTP) and early brain development. All of the NOS isoforms contain an N-terminal oxidase and a C-terminal reductase domain connected by a Ca(2+)/calmodulin binding region. To activate NOS I, Ca(2+) has to bind to calmodulin, allowing electron transport through both domains. Calcium ions are tightly regulated in cells. However, a number of other metal ions that bind and activate calmodulin may also activate NOS I. One such metal ion may be Pb(2+), which is associated with neurobehavioral and psychological alterations, including the inhibition of LTP. The effect of various divalent cations on NOS I activity was tested, and the results presented herein demonstrate that Pb(2+) and Sr(2+) can activate NOS I to a level similar to that found for Ca(2+). Finally, there is a synergy between Pb(2+) and Ca(2+) resulting in maximal activation of NOS I using minimal concentrations of both metal ions.
Sigma-Aldrich
Lead(II) carbonate, ACS reagent