MilliporeSigma
  • Tandem intermolecular Suzuki coupling/intramolecular vinyl triflate-arene coupling.
Tandem intermolecular Suzuki coupling/intramolecular vinyl triflate-arene coupling.
Chemical communications (Cambridge, England) (2002-07-19)
Michael C Willis, Christelle K Claverie, Mary F Mahon
ABSTRACT
Treatment of a benzyl substituted meso-ditriflate with boronic acids in the presence of palladium acetate, triphenylphosphine and caesium fluoride results in intermolecular Suzuki coupling followed by vinyl triflate-arene cyclisation to provide, in high yields, single regioisomers of tricyclic-carbocycles.
Sigma-Aldrich
Cesium fluoride, BioUltra, ≥99.0% (F)
Sigma-Aldrich
Cesium fluoride, purum p.a., ≥98.0%
Sigma-Aldrich
Cesium fluoride, 99.9% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Cesium fluoride, 99%
Sigma-Aldrich
Cesium Fluoride ChemBeads, NSC 84270