MilliporeSigma
  • Single-strand conformation polymorphism for p53 mutation by a combination of neutral pH buffer and temperature gradient in capillary electrophoresis.
Single-strand conformation polymorphism for p53 mutation by a combination of neutral pH buffer and temperature gradient in capillary electrophoresis.
Electrophoresis (2002-07-13)
Cecilia Gelfi, Agnese Vigano, Sara De Palma, Pier Giorgio Righetti, Sabin Carla Righetti, Elisabetta Corna, Franco Zunino
ABSTRACT
A large number of point mutations in the p53 gene have been detected by capillary zone electrophoresis via single-strand conformation polymorphism (SSCP) analysis. A much improved detection sensitivity was obtained via the following modifications in running conditions: use of low-viscosity 3% hydroxyethylcellulose (HEC), a neutral pH (pH 6.8) buffer, in which the standard Tris moiety was substituted with a 2-(N-morpholino)ethanesulfonic acid (MES)/Tris mixture, use of SYBR Green II for improved fluorescent signal at the lower pH adopted; and, finally, the use of a temperature gradient in the 15-25 degrees C interval, for favoring the conformational transitions in the mutated samples. The typical temperature gradient activated had a slope of 2 degrees C/min and were induced externally. A total of 24 samples from affected patients, both in the homo- and heterozygous state, were analyzed. All the mutations could be detected by this improved protocol, raising the sensitivity from the standard ca. 80% of conventional SSCP to essentially 100% with the present methodology. All the mutations were confirmed by sequence analysis of the affected samples.
Sigma-Aldrich
MES hydrate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
MES hydrate, ≥99.5% (titration), pH 2.5-4.0 (0.5 M in H2O), BioXtra
Sigma-Aldrich
MES hydrate, BioPerformance Certified, suitable for cell culture, ≥99.5%
Sigma-Aldrich
MES hydrate, ≥99.5% (titration)
Sigma-Aldrich
MES sodium salt, ≥99% (titration)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
MES sodium salt, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
MES sodium salt, BioReagent, ≥99% (titration), crystalline
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
SAFC
MES sodium salt