MilliporeSigma
  • Oxiranyl anion-mediated synthesis of highly enantiomerically enriched styrene oxide derivatives.
Oxiranyl anion-mediated synthesis of highly enantiomerically enriched styrene oxide derivatives.
Organic letters (2002-07-06)
Vito Capriati, Saverio Florio, Renzo Luisi, Antonio Salomone
ABSTRACT
[reaction: see text] The stereospecific alpha-lithiation of optically active styrene oxides and the trapping reaction of the corresponding highly reactive intermediates with electrophiles to produce optically active styrene oxide derivatives are described. This methodology has been applied to the synthesis of an optically active oral antifungal agent of industrial interest.
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric), tablet (5 g each)
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%