MilliporeSigma
  • [The effect of the dosage of TiO2 and its surface-loaded silver on the photocatalytic degradation of aroclor1260].
[The effect of the dosage of TiO2 and its surface-loaded silver on the photocatalytic degradation of aroclor1260].
Huan jing ke xue= Huanjing kexue (2002-06-07)
Chunlei Li, Bixian Mai, Haixiang Pan, Zheng Lin, Guoying Shen, Jiamo Fu
ABSTRACT
The dosage of TiO2 and its surface-loaded silver had notable effect on the photocatalytic degradation of Aroclor1260, under ultraviolet light. The dosage of TiO2 had no proportion to the degradation rate of Aroclor1260, but had an optimal value, which was 3 g/L under the experimental condition. The degradation rate of Aroclor1260 increased with the increasing of loading silver, and the PCBs congeners with higher concentration had higher degradation rate. The catalytic activity of TiO2 increased with the amount of loading silver increasing.
Product Description
Supelco
Aroclor 1260 solution, certified reference material, 500 mg/kg in transformer oil