MilliporeSigma
Distribution of fumonisins in dry-milled corn fractions in Argentina.
Food additives and contaminants (2002-05-25)
L E Broggi, S L Resnik, A M Pacin, H H L González, G Cano, D Taglieri
ABSTRACT
Corn samples and different dry-milled fractions collected from an industrial mill in Argentina were analysed. Average contaminations were FB(1) 1540 microg kg(-1), FB(2) 716 microg kg(-1) and FB(3) 152 microg kg(-1) in whole corn; FB(1) 135 microg kg(-1), FB(2) 39.1 microg kg(-1) and FB(3) 10.2 microg kg(-1) in corn grits; FB(1) 358 microg kg(-1), FB(2) 122 microg kg(-1) and FB(3) 45.9 microg kg(-1) in 'C' flour; FB(1) 148 microg kg(-1), FB(2) 52.5 microg kg(-1) and FB(3) 28.3 microg kg(-1) in corn meal; and FB(1) 4210 microg kg(-1), FB(2) 2010 microg kg(-1) and FB(3) 447 microg kg(-1) in germ and bran together. The fumonisin contamination level was approximately three times higher in germ and bran than in whole corn, 13 times higher than in 'C' flour and 29 times higher than in corn meal and corn grits. Taking into account the distribution of fumonisins in commercial dry-milled corn fractions and corn meal consumption in Argentina, a theoretical whole corn level of 6640 microg kg(-1) maximum of total fumonisins could be processed to obtain products considered safe for human health.
Product Description
Supelco
Fumonisin B3 solution, ~50 μg/mL in acetonitrile: water, analytical standard