MilliporeSigma
Kinetic resolutions of indan derivatives using bacteria.
Bioscience, biotechnology, and biochemistry (2002-05-10)
Naoki Tarui, Hayao Watanabe, Kohji Fukatsu, Shigenori Ohkawa, Kazuo Nakahama
ABSTRACT
Racemic indan derivatives have been resolved by the hydrolysis of amide bonds using Corynebacterium ammoniagenes IFO12612 to produce (S)-amine and (R)-amides. In the kinetic resolution of 1 (N-12-(6-methoxy-indan-1-yl)ethyl]acetamide), it was possible to run the reaction to 44% conversion on a 10-g scale, obtaining (S)-amine 4 ((S)-2-(6-methoxy-indan-1-yl)ethylamine) at >99% enantiomeric excess (ee) and (R)-1 at 98% ee.
Product Description
Supelco
Indan, analytical standard
Sigma-Aldrich
Indan, 95%