MilliporeSigma
  • Improved computed radiography image quality from a BaFl:Eu photostimulable phosphor plate.
Improved computed radiography image quality from a BaFl:Eu photostimulable phosphor plate.
Medical physics (2002-05-07)
Yasushi Nakano, Tomonori Gido, Satoshi Honda, Akihiro Maezawa, Hideaki Wakamatsu, Takafumi Yanagita
ABSTRACT
Recent advances in the design of photostimulable phosphor (PSP) plates for computed radiography (CR) systems have made it possible to manufacture plates made of BaFI:Eu phosphor material in the cassette form. The image quality of this plate, six BaF(Br,I):Eu plates, and one BaFBr:Eu plate were evaluated in terms of presampling modulation transfer function (MTF), normalized Wiener spectra (WSN), and detective quantum efficiency (DQE). Compared with the best-performing BaF(Br,I):Eu plate, the BaFI:Eu plate provided DQE that was higher, at spatial frequencies of 0.5, 1.0, and 2.0 cycles/mm, by 12% (21.8 versus 19.4), 13% (18.8 versus 16.7), and 11% (12.0 versus 10.8), respectively. Since presampling MTF values of the two plates were comparable, the BaFI:Eu plate's higher DQE is attributable to total WSN conversely being lower by 17% [8.65 x 10(-5) (mm2) versus 10.38 x 10(-5) (mm2)], 17% [5.85 x 10(-5) (mm2) versus 7.03 x 10(-5) (mm2)], and 12% [2.82 x 10(-5) (mm2) versus 3.19 x 10(-5) (mm2)] at the specified frequencies, respectively, primarily due to the contribution of x-ray quantum WSN. This jibes with the high x-ray absorption provided by the 27%-higher x-ray attenuation coefficients (7.54 versus 6.07, at 60 KeV) that BaFI offers over BaF(Br(0.85),I(0.15)), a result of the high atomic number of BaFI's exclusively iodine halide content. The results were consistent with earlier studies of several of these same plates and indicate that BaFI:Eu is a promising avenue to lower image noise and higher overall CR system image quality.
Sigma-Aldrich
Barium fluoride, precipitated, 99.95% trace metals basis
Sigma-Aldrich
Barium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Barium fluoride, powder, 98%