MilliporeSigma
  • Hydroxyl radical formation and oxidative DNA damage induced by areca quid in vivo.
Hydroxyl radical formation and oxidative DNA damage induced by areca quid in vivo.
Journal of toxicology and environmental health. Part A (2002-03-26)
Chiu-Lan Chen, Chin-Wen Chi, Tsung-Yun Liu
ABSTRACT
Chewing areca quid (AQ) has been implicated as a major risk factor for the development of oral squamous-cell carcinoma (OSCC). Recent studies have suggested that AQ-generated reactive oxygen species (ROS) is one of the contributing factors for oral carcinogenesis. However, the AQ used in Taiwan is different from that used in other countries. This study is designed to test whether ROS are generated and the consequent effects in locally prepared AQ in vivo. We measured the hydroxyl radical formation, as represented by the presence of o- and m-tyrosine in saliva from volunteers who chewed AQ containing 20 mg phenylalanine. Their saliva contained significantly higher amounts (p < .05) of o- and m-tyrosine as compared to the controls. In addition, chewing AQ containing Piper betle inflorescence generated higher amounts of m-tyrosine, but not o-tyrosine, in saliva than did chewing AQ containing betel leaf. We further tested the oxidative DNA damaging effect of the reconstituted AQ, as evidenced by the elevation of 8-hydroxy-2'-deoxyguanosine (8-OH-dG) levels, in hamster buccal pouch. Following daily painting for 14 d, the 8-OH-dG level in hamster buccal pouch is significantly elevated (p < .05) in the AQ-treated group versus the controls. These findings demonstrate that ROS, such as hydroxyl radical, are formed in the human oral cavity during AQ chewing, and chewing such prepared AQ might cause oxidative DNA damage to the surrounding tissues.
Sigma-Aldrich
DL-m-Tyrosine, crystalline