MilliporeSigma
  • Lipase-catalyzed synthesis of a pure macrocyclic polyester from dimethyl terephthalate and diethylene glycol.
Lipase-catalyzed synthesis of a pure macrocyclic polyester from dimethyl terephthalate and diethylene glycol.
Biomacromolecules (2002-03-13)
Arnaud Lavalette, Thierry Lalot, Maryvonne Brigodiot, Ernest Maréchal
ABSTRACT
The polymerization of dimethyl terephthalate and diethylene glycol by enzymic catalysis in toluene is described. The potential pi-pi interactions of the aromatic rings together with the relative flexibility of the diol segment have been regarded as synergy factors responsible for the formation of a pure cyclic compound. The so-called C2 macrocycle was characterized by (1)H and (13)C NMR, size-exclusion chromatography, differential scanning calorimetry, mass spectrometry, and X-ray diffraction on powder. Structurally speaking, it is made of two repeating units. The substitution of the central oxygen atom of the diol by -CH(2)- or -S- as well as the presence of ortho-substituents (-NH(2) or -NO(2)) on the phthalate were expected to disrupt the stacking of the phenyl groups more or less.
Sigma-Aldrich
Dimethyl terephthalate, purum, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl terephthalate, ReagentPlus®, ≥99%
Supelco
Dimethyl terephthalate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland