MilliporeSigma
  • Distribution of triazolam and alpha-hydroxytriazolam in a fatal intoxication case.
Distribution of triazolam and alpha-hydroxytriazolam in a fatal intoxication case.
Journal of analytical toxicology (2002-03-13)
Barry Levine, Alison Grieshaber, Joseph Pestaner, Karla A Moore, John E Smialek
ABSTRACT
The case of a 77-year-old woman who was found dead in her bathtub with her head clearly above the water line is presented. The decedent had a medical history of depression, liver disease, spinal stenosis, and diabetes mellitus. An empty medication bottle of triazolam was found in the trashcan. At autopsy, no injury or evidence of drowning was found. Toxicological analysis identified triazolam at a concentration of 0.12 mg/L in the heart blood. Triazolam and alpha-hydroxytriazolam were quantitated in the specimens received. The medical examiner ruled that the cause of death was triazolam intoxication and the manner of death was suicide.
Product Description
Supelco
α-Hydroxytriazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®