MilliporeSigma
Synthesis of the sialidase inhibitor siastatin B.
Organic letters (2000-12-12)
S Knapp, D Zhao
ABSTRACT
[structure] The resolved piperidinecarboxylate (R)-7 was converted to siastatin B (1) by an efficient and stereoselective sequence that includes a bromo-beta-lactonization and an N-acyliminium azidation. Two analogues (3 and 4) of siastatin were also prepared.
Sigma-Aldrich
Siastatin B, lyophilized powder, from microbial