MilliporeSigma
  • Isolation and structure elucidation of a new prenylcoumarin from Murraya paniculata var. omphalocarpa (Rutaceae).
Isolation and structure elucidation of a new prenylcoumarin from Murraya paniculata var. omphalocarpa (Rutaceae).
Chemical & pharmaceutical bulletin (2002-02-05)
Takeshi Kinoshita, Motoko Shimada
ABSTRACT
A new C-8 prenylated 5,7-dimethoxycoumarin named omphamurrayin was isolated from the leaves of Murraya paniculata var. omphalocarpa, and its structure was established as 5,7-dimethoxy-8-(1-oxo-2-senecioyl-3-methyl-3-butenyl)-2H-1-benzopyran-2-one on the basis of the spectroscopic evidence. The taxonomic status of M. paniculata var. omphalocarpa is briefly discussed, along with its synonymity to M. paniculata from the chemosystematic viewpoint.
Sigma-Aldrich
5,7-Dimethoxycoumarin, 98%